siRNA Details
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021363.2 | NP_067338.2 | 1, 2 | 105 | ||
GenBank mRNA | AF242220.1 | - | 1, 2 | 90 | ||
BC131996.1 | - | 1, 2 | 105 | 40130273 | ||
BC131998.1 | - | 1, 2 | 105 | 40130275 |
Gene Symbol: Svs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173377.2 | NP_775553.1 | 1, 2 | 106 | ||
GenBank mRNA | AK035235.1 | - | 1, 2 | 104 | ||
BY009043.1 | - | 1, 2 | 107 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
