siRNA Details
Gene Symbol: Xlr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293676.1 | NP_001280605.1 | 3, 4 | 355 | ||
NM_001293677.1 | NP_001280606.1 | 3, 4 | 308 | |||
NM_001293678.1 | NP_001280607.1 | 3, 4 | 332 | |||
NM_001293679.1 | NP_001280608.1 | 3, 4 | 344 | |||
NM_001293680.1 | NP_001280609.1 | 3, 4 | 368 | |||
NM_021365.3 | NP_067340.2 | 3, 4 | 472 | |||
GenBank mRNA | AL136330.1 | - | 3, 4 | 440 | ||
BC025576.1 | - | 3, 4 | 308 | 5323621 | ||
BC034848.1 | - | 3, 4 | 896 | |||
BC138975.1 | - | 3, 4 | 214 | 8861997 | ||
BC138979.1 | - | 3, 4 | 238 | 8862001 | ||
BC145481.1 | - | 3, 4 | 238 | 9054044 | ||
BC145482.1 | - | 3, 4 | 250 | 9054045 | ||
BC145483.1 | - | 3, 4 | 274 | 9054046 | ||
BE627633.1 | - | 3, 4 | 271 |
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081642.1 | NP_001075111.1 | 3, 4 | 344 | ||
GenBank mRNA | AK041345.1 | - | 1 | 281 | ||
BC098242.1 | - | 3, 4 | 324 | 1478336 |
Gene Symbol: Xlr4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183094.3 | NP_898917.2 | 3, 4 | 374 | ||
GenBank mRNA | AK013623.1 | - | 3, 4 | 259 | ||
AK166871.1 | - | 3, 4 | 375 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
