siRNA Details
Gene Symbol: Prg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110146.1 | NP_001103616.1 | 3 | 157 | ||
NM_021400.3 | NP_067375.3 | 3 | 157 | |||
GenBank mRNA | AB034730.1 | - | 3 | 157 | ||
AK149469.1 | - | 3 | 158 | |||
BC130021.1 | - | 2 | 121 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
