siRNA Details
Gene Symbol: Gsdma2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029727.2 | NP_084003.2 | 4, 5 | 535 | ||
GenBank mRNA | AK008663.1 | - | 4, 5 | 532 | ||
BC109335.1 | - | 3, 4 | 438 | 40052139 | ||
BC109336.1 | - | 4, 5 | 469 | 40052146 |
Gene Symbol: Gsdma | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021347.4 | NP_067322.1 | 3, 4 | 496 | ||
GenBank mRNA | AB033595.1 | - | 3, 4 | 485 | ||
AK028645.1 | - | 3, 4 | 497 | |||
AK028698.1 | - | 3, 4 | 497 | |||
AK029062.1 | - | 3, 4 | 480 |
Gene Symbol: Gsdma3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007461.1 | NP_001007462.1 | 3, 4 | 485 | ||
GenBank mRNA | AY679090.1 | - | 3, 4 | 485 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
