siRNA Details
Gene Symbol: Xlr4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293676.1 | NP_001280605.1 | 4, 5 | 424 | ||
NM_001293677.1 | NP_001280606.1 | 4, 5 | 377 | |||
NM_001293678.1 | NP_001280607.1 | 4, 5 | 401 | |||
NM_001293679.1 | NP_001280608.1 | 4, 5 | 413 | |||
NM_001293680.1 | NP_001280609.1 | 4, 5 | 437 | |||
NM_021365.3 | NP_067340.2 | 4, 5 | 541 | |||
GenBank mRNA | AL136330.1 | - | 4, 5 | 509 | ||
BC025576.1 | - | 4, 5 | 377 | 5323621 | ||
BC034848.1 | - | 4, 5 | 965 | |||
BC138975.1 | - | 4, 5 | 283 | 8861997 | ||
BC138979.1 | - | 4, 5 | 307 | 8862001 | ||
BC145481.1 | - | 4, 5 | 307 | 9054044 | ||
BC145482.1 | - | 4, 5 | 319 | 9054045 | ||
BC145483.1 | - | 4, 5 | 343 | 9054046 | ||
BE627633.1 | - | 4, 5 | 340 |
Gene Symbol: Xlr4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081642.1 | NP_001075111.1 | 4, 5 | 413 | ||
GenBank mRNA | BC098242.1 | - | 4, 5 | 393 | 1478336 |
Gene Symbol: Xlr4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183094.3 | NP_898917.2 | 4, 5 | 443 | ||
GenBank mRNA | AK013623.1 | - | 4, 5 | 328 | ||
AK166871.1 | - | 4, 5 | 444 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
