siRNA Details
Gene Symbol: Rhox4c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039689.1 | NP_001034778.1 | 4 | 712 | ||
GenBank mRNA | AJ972667.1 | - | 4 | 712 |
Gene Symbol: Rhox4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039688.3 | NP_001034777.1 | 4 | 700 | ||
GenBank mRNA | AJ972665.1 | - | 4 | 700 | ||
AK017380.1 | - | 4 | 615 | |||
BC057925.1 | - | 4 | 615 | 5292192 | ||
BC116370.1 | - | 4 | 648 | 40100097 | ||
BC116371.1 | - | 4 | 647 | 40100098 | ||
BC145382.1 | - | 4 | 601 | 9053940 | ||
BC171959.1 | - | 4 | 601 | 9054245 | ||
DQ058641.1 | - | 4 | 675 |
Gene Symbol: Rhox4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021300.2 | NP_067275.1 | 4 | 695 | ||
GenBank mRNA | AF265350.1 | - | 4 | 683 | ||
AJ972666.1 | - | 4 | 695 | |||
BC172126.1 | - | 4 | 642 | 9054796 |
Gene Symbol: Rhox4d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039695.1 | NP_001034784.1 | 568 | |||
GenBank mRNA | AB221641.1 | - | 564 | |||
AJ972668.1 | - | 568 | ||||
BC147696.1 | - | 4 | 642 | 9056669 |
Gene Symbol: Rhox4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_201236.3 | NP_957688.2 | 4 | 786 | ||
GenBank mRNA | AJ972669.1 | - | 4 | 786 | ||
BC094891.1 | - | 4 | 748 | 30019143 | ||
BC172128.1 | - | 642 | 9054798 |
Gene Symbol: Rhox4f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039696.1 | NP_001034785.1 | 4 | 738 | ||
GenBank mRNA | AJ972670.1 | - | 4 | 738 |
Gene Symbol: Rhox4g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039698.1 | NP_001034787.1 | 4 | 787 | ||
GenBank mRNA | AJ972671.1 | - | 4 | 787 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
