siRNA Details
Gene Symbol: Noc2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021303.2 | NP_067278.2 | 18 | 2155 | ||
GenBank mRNA | AF155546.1 | - | 6 | 665 | ||
AK076287.1 | - | 17 | 2197 | |||
AK157765.1 | - | 18 | 2148 | |||
AK199140.1 | - | 2 | 158 | |||
BC020013.1 | - | 18 | 2126 | 3663482 |
