siRNA Details
Gene Symbol: Sh2d2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021309.3 | NP_067284.1 | 6 | 1072 | ||
GenBank mRNA | AF203343.1 | - | 6 | 1079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
