siRNA Details
Gene Symbol: Fhl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021318.3 | NP_067293.1 | 5 | 786 | ||
GenBank mRNA | AF083394.1 | - | 5 | 734 | ||
AK006414.1 | - | 5 | 775 | |||
BC048473.1 | - | 5 | 786 | 6770515 |

More Information
Other Species Targeted

Gene Ontology Categories:
