siRNA Details
Gene Symbol: Svs3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173377.2 | NP_775553.1 | 2 | 118 | ||
GenBank mRNA | AK035235.1 | - | 2 | 116 |
Gene Symbol: Svs3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021363.2 | NP_067338.2 | 2 | 117 | ||
GenBank mRNA | AF242220.1 | - | 2 | 102 | ||
AK020575.1 | - | 2 | 113 | |||
BC131996.1 | - | 2 | 117 | 40130273 | ||
BC131998.1 | - | 2 | 117 | 40130275 | ||
BY722061.1 | - | 2 | 113 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
