siRNA Details
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153840.3 | NP_722582.2 | 8 | 923 | ||
XM_011514469.1 | XP_011512771.1 | 5 | 579 | |||
XM_017010735.1 | XP_016866224.1 | 5 | 681 | |||
XM_017010736.1 | XP_016866225.1 | 6 | 656 | |||
GenBank mRNA | AB045123.1 | - | 8 | 877 | ||
AK095348.1 | - | 6 | 740 | |||
AY140952.1 | - | 2 | 116 | |||
BC029358.1 | - | 7 | 748 | |||
CR627234.1 | - | 7 | 4487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Set Membership: |
![]() |
Gene Ontology Categories:
