siRNA Details
Gene Symbol: Ush2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021408.3 | NP_067383.3 | 1 | 345 | ||
GenBank mRNA | AF151717.1 | - | 2 | 374 | ||
DQ073638.1 | - | 1 | 149 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
