siRNA Details
Gene Symbol: DDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014796.1 | NP_001014796.1 | 5 | 585 | ||
NM_006182.2 | NP_006173.2 | 4 | 505 | |||
XM_006711344.3 | XP_006711407.1 | 4 | 546 | |||
XM_011509586.2 | XP_011507888.1 | 4 | 478 | |||
XM_011509587.2 | XP_011507889.1 | 5 | 539 | |||
XM_011509588.2 | XP_011507890.1 | 4 | 542 | |||
GenBank mRNA | AK095975.1 | - | 5 | 558 | ||
AK225063.1 | - | 4 | 424 | |||
AK225196.1 | - | 4 | 480 | |||
AK314388.1 | - | 4 | 454 | |||
BC052998.2 | - | 5 | 556 | 6162716 | ||
BX640899.1 | - | 5 | 542 | |||
CF994640.1 | - | 4 | 555 | |||
X74764.1 | - | 4 | 473 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
