siRNA Details
Gene Symbol: Tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021480.5 | NP_067455.5 | 3, 4 | 634 | ||
GenBank mRNA | AF134346.2 | - | 3, 4 | 298 | ||
AK077571.1 | - | 3, 4 | 309 | |||
AY116662.1 | - | 2, 3 | 235 |

More Information
Other Species Targeted

Gene Ontology Categories:
