siRNA Details
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033621.3 | NP_001028793.1 | 2 | 531 | ||
GenBank mRNA | AF230979.2 | - | 2 | 530 | ||
AK017447.1 | - | 2 | 532 | |||
BC016214.1 | - | 2 | 509 | 4482382 | ||
BC064052.1 | - | 2 | 504 | 1366418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
