siRNA Details
Gene Symbol: Stx6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021433.3 | NP_067408.1 | 2 | 384 | ||
GenBank mRNA | AF237814.1 | - | 2 | 194 | ||
AK009690.1 | - | 2 | 339 | |||
AK018167.1 | - | 2 | 372 | |||
AK019106.1 | - | 2 | 346 | |||
AK031673.1 | - | 2 | 363 | |||
AK044905.1 | - | 2 | 345 | |||
AK049124.1 | - | 2 | 344 | |||
AK151549.1 | - | 2 | 338 | |||
AK159392.1 | - | 2 | 356 | |||
BC029205.1 | - | 2 | 301 | 4039560 | ||
BQ749205.1 | - | 2 | 357 |
