siRNA Details
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229032.1 | - | 3 | 257 | ||
BC033458.1 | - | 3 | 263 | |||
CD766233.1 | - | 3 | 307 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
