siRNA Details
Gene Symbol: Trpm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164325.1 | NP_001157797.1 | 4 | 512 | ||
NM_021450.2 | NP_067425.2 | 4 | 512 | |||
GenBank mRNA | AF149013.1 | - | 4 | 462 | ||
AF376052.1 | - | 4 | 368 | |||
AK014698.1 | - | 4 | 453 | |||
AK052823.1 | - | 4 | 484 | |||
AK162483.1 | - | 4 | 438 | |||
AY032951.1 | - | 4 | 481 | |||
BC145122.1 | - | 4 | 307 | |||
BC150709.1 | - | 4 | 307 | 9087620 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_053705.
Gene Ontology Categories:
