siRNA Details
Gene Symbol: Prps1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021463.4 | NP_067438.1 | 1, 2 | 261 | ||
GenBank mRNA | AB025048.1 | - | 1, 2 | 191 | ||
AK009265.1 | - | 1, 2 | 239 | |||
AK011034.1 | - | 1, 2 | 239 | |||
AK011304.1 | - | 1, 2 | 240 | |||
AK089009.1 | - | 1, 2 | 256 | |||
AK143609.1 | - | 1, 2 | 239 | |||
AK165524.1 | - | 1, 2 | 240 | |||
AK165987.1 | - | 1, 2 | 242 | |||
AK166856.1 | - | 1, 2 | 240 | |||
AK168036.1 | - | 1, 2 | 173 | |||
AK185462.1 | - | 1, 2 | 59 | |||
BC054772.1 | - | 1, 2 | 212 | 6415720 | ||
BY003422.1 | - | 1, 2 | 261 |
Gene Symbol: Prps1l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037746.3 | NP_001032835.2 | 2 | 385 | ||
GenBank mRNA | AK134500.1 | - | 2 | 386 | ||
BC110357.1 | - | 2 | 288 | 5293637 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
