siRNA Details
Gene Symbol: Tnni1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112702.1 | NP_001106173.1 | 7 | 319 | ||
NM_021467.5 | NP_067442.1 | 7 | 367 | |||
GenBank mRNA | AA230582.1 | - | 7 | 367 | ||
AA271187.1 | - | 7 | 319 | |||
AJ242874.1 | - | 7 | 367 | |||
AK012258.1 | - | 7 | 350 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
