siRNA Details
Gene Symbol: 0610007P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021446.2 | NP_067421.1 | 5 | 1082 | ||
GenBank mRNA | AF270646.1 | - | 5 | 1082 | ||
AK002308.1 | - | 4 | 1082 | |||
AK004480.1 | - | 5 | 1104 | |||
AK152230.1 | - | 5 | 1060 | |||
BC004591.1 | - | 5 | 1068 | 3587693 |
