siRNA Details
Gene Symbol: Crbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021449.3 | NP_067424.2 | 11 | 1749 | ||
NM_175357.3 | NP_780566.1 | 11 | 1752 | |||
GenBank mRNA | AF229032.1 | - | 11 | 1690 | ||
AK076144.1 | - | 11 | 1732 | |||
AK077707.1 | - | 11 | 1732 | |||
AK160219.1 | - | 11 | 1728 | |||
BC046967.1 | - | 10 | 1667 | |||
BC069905.1 | - | 11 | 1709 | |||
BC086488.1 | - | 11 | 1727 | 30443656 |

More Information
Other Species Targeted

Gene Ontology Categories:
