siRNA Details
Gene Symbol: Rnase4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB041044.1 | - | 2 | 901 | ||
BC005569.1 | - | 2 | 869 | 3967265 | ||
BC066843.1 | - | 2 | 956 | 30059308 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
