siRNA Details
Gene Symbol: Myot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033621.3 | NP_001028793.1 | 10 | 1684 | ||
GenBank mRNA | AF230979.2 | - | 10 | 1683 | ||
AK017447.1 | - | 10 | 1685 | |||
BC016214.1 | - | 10 | 1662 | 4482382 | ||
BC064052.1 | - | 10 | 1657 | 1366418 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
