siRNA Details
Gene Symbol: Chst12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021528.3 | NP_067503.3 | 2 | 343 | ||
GenBank mRNA | AJ289132.1 | - | 1 | 256 | ||
AK142155.1 | - | 2 | 341 | |||
AK170209.1 | - | 2 | 306 | |||
AK170484.1 | - | 2 | 310 | |||
AK171208.1 | - | 2 | 310 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
