siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 2, 3 | 318 | ||
GenBank mRNA | AF224508.1 | - | 2, 3 | 264 | ||
AK034237.1 | - | 2, 3 | 320 | |||
AK039404.1 | - | 2, 3 | 283 | |||
AK173014.1 | - | 2, 3 | 152 | |||
BB626223.1 | - | 2, 3 | 319 | |||
BB851903.1 | - | 2, 3 | 316 | |||
BC030388.1 | - | 2, 3 | 280 | 5401143 |

More Information
Other Species Targeted

Gene Ontology Categories:
