siRNA Details
Gene Symbol: Hist1h3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013550.5 | NP_038578.2 | 1 | 445 | ||
GenBank mRNA | AK006742.1 | - | 1 | 425 | ||
AK018952.1 | - | 1 | 446 | |||
BC115816.1 | - | 1 | 442 | 40105082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
