siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 4 | 509 | ||
GenBank mRNA | AF224508.1 | - | 4 | 455 | ||
AK034237.1 | - | 4 | 511 | |||
AK039404.1 | - | 4 | 474 | |||
AK171610.1 | - | 3 | 285 | |||
AK173014.1 | - | 4 | 343 | |||
BB626223.1 | - | 4 | 510 | |||
BC030388.1 | - | 4 | 471 | 5401143 |
