siRNA Details
Gene Symbol: Slc22a17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB012808.1 | - | 7, 8 | 1006 | ||
BC005727.1 | - | 5, 6 | 1120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
