siRNA Details
Gene Symbol: Bhlhe22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021560.4 | NP_067535.3 | 1 | 1666 | ||
GenBank mRNA | AF218865.1 | - | 1 | 1288 | ||
AK047201.1 | - | 1 | 1400 | |||
BC053007.1 | - | 1 | 1666 | 5686844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
