siRNA Details
Gene Symbol: Slc4a8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021530.2 | NP_067505.2 | 18 | 2490 | ||
GenBank mRNA | AF224508.1 | - | 18 | 2436 | ||
AK034237.1 | - | 18 | 2492 | |||
AK039404.1 | - | 18 | 2455 | |||
AK047863.1 | - | 11 | 1424 | |||
AK171610.1 | - | 17 | 2266 | |||
AK173014.1 | - | 18 | 2324 | |||
AK183206.1 | - | 3 | 236 | |||
BC030388.1 | - | 18 | 2452 | 5401143 |
