siRNA Details
Gene Symbol: Hgh1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021555.2 | NP_067530.2 | 6 | 1885 | ||
GenBank mRNA | AB041552.1 | - | 6 | 1886 | ||
AK085815.1 | - | 6 | 1879 | |||
BC061251.1 | - | 6 | 1849 | 30249656 |
Gene Symbol: Dclre1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK046571.1 | - | 3 | 1344 | ||
AK163827.1 | - | 2 | 1942 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
