siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022027.2 | NP_071310.2 | 1, 2 | 265 | ||
GenBank mRNA | AF281869.1 | - | 1, 2 | 181 | ||
AK036828.1 | - | 1, 2 | 252 | |||
BC054456.1 | - | 1, 2 | 265 | |||
BC110428.1 | - | 1, 2 | 245 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
