siRNA Details
Gene Symbol: Oxct2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181859.3 | NP_862907.2 | 1 | 298 | ||
GenBank mRNA | AB049996.1 | - | 1 | 288 | ||
BC113758.1 | - | 1 | 281 | 40047052 | ||
CJ047771.1 | - | 1 | 299 |
Gene Symbol: Oxct2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022033.4 | NP_071316.1 | 1 | 350 | ||
GenBank mRNA | AB022180.2 | - | 1 | 288 | ||
AK077075.1 | - | 1 | 303 | |||
BC137887.1 | - | 1 | 350 | 8860906 | ||
BC137888.1 | - | 1 | 350 | 8860907 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
