siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079686.1 | NP_001073154.1 | 40 | 6455 | ||
NM_022027.2 | NP_071310.2 | 2 | 409 | |||
GenBank mRNA | AF281869.1 | - | 2 | 325 | ||
AF281870.1 | - | 21 | 3366 | |||
AK036828.1 | - | 2 | 396 | |||
BC054456.1 | - | 2 | 409 | |||
BC110426.1 | - | 11 | 1717 | |||
BC110428.1 | - | 2 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
