siRNA Details
Gene Symbol: Ctsz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022325.4 | NP_071720.1 | 5 | 816 | ||
GenBank mRNA | AF136277.1 | - | 5 | 826 | ||
AF197479.1 | - | 5 | 743 | |||
AJ242663.1 | - | 5 | 826 | |||
AK002710.1 | - | 5 | 778 | |||
AK004095.1 | - | 5 | 814 | |||
AK008370.1 | - | 5 | 745 | |||
AK010912.1 | - | 5 | 812 | |||
AK150329.1 | - | 5 | 744 | |||
AK150894.1 | - | 5 | 751 | |||
AK150947.1 | - | 5 | 805 | |||
AK151440.1 | - | 5 | 755 | |||
AK151585.1 | - | 5 | 779 | |||
AK151994.1 | - | 5 | 789 | |||
AK153147.1 | - | 5 | 751 | |||
AK159118.1 | - | 5 | 762 | |||
AK159618.1 | - | 5 | 829 | |||
AK159826.1 | - | 5 | 751 | |||
AK160001.1 | - | 5 | 799 | |||
AK160028.1 | - | 5 | 799 | |||
AK160354.1 | - | 5 | 832 | |||
AK166047.1 | - | 5 | 800 | |||
AK185170.1 | - | 1 | 75 | |||
AK185852.1 | - | 1 | 75 | |||
AK186484.1 | - | 2 | 126 | |||
AK192329.1 | - | 1 | 75 | |||
AK204750.1 | - | 1 | 17 | |||
AK205915.1 | - | 1 | 20 | |||
AK215212.1 | - | 1 | 75 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
