siRNA Details
Gene Symbol: Rnf187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022423.2 | NP_071868.2 | 2 | 549 | ||
GenBank mRNA | AB030190.1 | - | 2 | 541 | ||
AK037991.1 | - | 2 | 464 | |||
AK075944.1 | - | 2 | 560 | |||
AK076158.1 | - | 2 | 539 | |||
AK088973.1 | - | 2 | 552 | |||
BC072613.1 | - | 2 | 542 | |||
BC096522.1 | - | 2 | 513 |

More Information
Other Species Targeted

Gene Ontology Categories:
