siRNA Details
Gene Symbol: Syne1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079686.1 | NP_001073154.1 | 51 | 8390 | ||
NM_022027.2 | NP_071310.2 | 13 | 2344 | |||
GenBank mRNA | AF281869.1 | - | 13 | 2260 | ||
AF281870.1 | - | 32 | 5301 | |||
AK036828.1 | - | 14 | 2400 | |||
BC041779.1 | - | 12 | 1941 | |||
BC054456.1 | - | 13 | 2344 | |||
BC110426.1 | - | 22 | 3652 | |||
BC110428.1 | - | 13 | 2324 | |||
BC111034.1 | - | 2 | 243 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_006227852, XM_006227851, XM_008758713, AF452647.
Gene Ontology Categories:
