siRNA Details
Gene Symbol: Myh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022410.3 | NP_071855.2 | 41 | 7235 | ||
GenBank mRNA | AJ312390.1 | - | 41 | 7158 | ||
AK087164.1 | - | 1 | 686 | |||
AK131171.1 | - | 27 | 5058 | |||
AK147203.1 | - | 41 | 7158 | |||
AK147206.1 | - | 41 | 7167 | |||
AK147208.1 | - | 41 | 7175 | |||
AK147209.1 | - | 41 | 7166 | |||
AK147210.1 | - | 41 | 7166 | |||
AK147211.1 | - | 41 | 7210 | |||
AK147215.1 | - | 41 | 7153 | |||
AK147216.1 | - | 41 | 7108 | |||
AK147221.1 | - | 41 | 7113 | |||
AK147222.1 | - | 41 | 7154 | |||
AK147223.1 | - | 41 | 7159 | |||
AK147233.1 | - | 41 | 7166 | |||
AK147235.1 | - | 41 | 7166 | |||
AK147296.1 | - | 41 | 7166 | |||
AK147407.1 | - | 41 | 7169 | |||
AK147430.1 | - | 41 | 7176 | |||
BC026521.1 | - | 1 | 798 | |||
BC054360.1 | - | 9 | 2253 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
