siRNA Details
Gene Symbol: Rnf187 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022423.2 | NP_071868.2 | 2 | 553 | ||
GenBank mRNA | AB030190.1 | - | 2 | 545 | ||
AK037991.1 | - | 2 | 468 | |||
AK075944.1 | - | 2 | 564 | |||
AK076158.1 | - | 2 | 543 | |||
AK088973.1 | - | 2 | 556 | |||
BC072613.1 | - | 2 | 546 | |||
BC096522.1 | - | 2 | 517 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
