siRNA Details
Gene Symbol: Kng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102411.1 | NP_001095881.1 | 1 | 245 | ||
NM_001102412.1 | NP_001095882.1 | 1 | 245 | |||
NM_023125.3 | NP_075614.1 | 1 | 245 | |||
GenBank mRNA | AK005547.1 | - | 1 | 210 | ||
AY462058.1 | - | 1 | 125 | |||
AY660571.1 | - | 1 | 152 | |||
BC018158.1 | - | 1 | 197 | 4162960 | ||
BC108936.1 | - | 1 | 201 | 40039851 | ||
D84415.1 | - | 1 | 125 | |||
D84435.1 | - | 1 | 125 |
Gene Symbol: Kng2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102409.1 | NP_001095879.1 | 1 | 293 | ||
NM_001102410.1 | NP_001095880.1 | 1 | 293 | |||
NM_201375.2 | NP_958763.1 | 1 | 293 | |||
GenBank mRNA | AY462056.1 | - | 1 | 209 | ||
AY462057.1 | - | 1 | 209 | |||
AY462059.1 | - | 1 | 209 | |||
AY660572.1 | - | 1 | 209 | |||
BC108335.1 | - | 1 | 260 | 5124483 | ||
BC108934.1 | - | 1 | 201 | |||
BC139720.1 | - | 1 | 201 | 40039854 | ||
BY773346.1 | - | 1 | 294 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK128967.1 | - | 124 | Chr. - - [-] on Build |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
