siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164207.1 | NP_001157679.1 | 5 | 832 | ||
NM_001164208.1 | NP_001157680.1 | 5 | 480 | |||
NM_001164209.2 | NP_001157681.1 | 6 | 688 | |||
NM_001286651.1 | NP_001273580.1 | 6 | 685 | |||
NM_001286652.1 | NP_001273581.1 | 5 | 592 | |||
NM_023056.4 | NP_075543.1 | 5 | 1081 | |||
GenBank mRNA | AB031386.1 | - | 5 | 641 | ||
AF115426.1 | - | 5 | 553 | |||
AK156042.1 | - | 5 | 833 | |||
BC019539.1 | - | 4 | 450 | 4237639 | ||
BC022166.1 | - | 5 | 1096 | 3582123 | ||
BQ571258.1 | - | 5 | 472 | |||
CT010176.1 | - | 4 | 445 | |||
DV047496.1 | - | 5 | 520 | |||
DV059241.1 | - | 5 | 556 |
