siRNA Details
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023119.2 | NP_075608.2 | 4 | 353 | ||
GenBank mRNA | AK002322.1 | - | 4 | 312 | ||
AK002336.1 | - | 4 | 311 | |||
AK014335.1 | - | 3 | 1292 | |||
AK088788.1 | - | 4 | 297 | |||
AK147987.1 | - | 4 | 311 | |||
AK150161.1 | - | 4 | 311 | |||
AK150192.1 | - | 4 | 312 | |||
AK153321.1 | - | 4 | 310 | |||
AK180361.1 | - | 2 | 35 | |||
AK191162.1 | - | 1 | 35 | |||
AK191332.1 | - | 2 | 35 | |||
AK193395.1 | - | 35 | ||||
AK196981.1 | - | 2 | 35 | |||
AK199229.1 | - | 2 | 35 | |||
AK202164.1 | - | 2 | 35 | |||
AK202428.1 | - | 2 | 35 | |||
AK203149.1 | - | 2 | 35 | |||
AK204087.1 | - | 2 | 35 | |||
AK204457.1 | - | 2 | 35 | |||
AK204949.1 | - | 2 | 35 | |||
AK214077.1 | - | 3 | 169 | |||
BC003891.1 | - | 4 | 303 | 3592683 | ||
BC003934.1 | - | 4 | 325 | |||
BC010685.1 | - | 4 | 291 | 3599051 | ||
BC024644.1 | - | 4 | 283 | 3583123 | ||
BC039179.1 | - | 4 | 273 | 4501041 | ||
BC083334.1 | - | 4 | 285 | 6414729 | ||
BC085098.1 | - | 3 | 214 | 5376359 | ||
BU526382.1 | - | 4 | 356 | |||
X52379.1 | - | 4 | 295 |
Gene Symbol: Eno1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025388.1 | NP_001020559.1 | 2 | 335 | ||
GenBank mRNA | BC089539.1 | - | 2 | 335 | 6707317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
