siRNA Details
Gene Symbol: Palm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161747.1 | NP_001155219.1 | 4 | 376 | ||
NM_023128.4 | NP_075617.3 | 4 | 376 | |||
GenBank mRNA | AK009200.1 | - | 3 | 217 | ||
AK082163.1 | - | 4 | 378 | |||
AK129107.1 | - | 2 | 115 | |||
AK133733.1 | - | 4 | 282 | |||
AK162411.1 | - | 4 | 369 | |||
BC015297.1 | - | 4 | 360 | 4223845 | ||
Y14771.1 | - | 4 | 339 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
