siRNA Details
Gene Symbol: Lrp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022993.3 | NP_075369.2 | 7 | 2500 | ||
GenBank mRNA | AB042200.1 | - | 7 | 2402 | ||
AK132169.1 | - | 2484 | ||||
AK144035.1 | - | 7 | 2513 | |||
AK170325.1 | - | 7 | 2474 | |||
AK193662.1 | - | 1 | 92 | |||
AK196385.1 | - | 1 | 92 | |||
AK199366.1 | - | 1 | 92 | |||
AK206255.1 | - | 1 | 92 | |||
AK210018.1 | - | 1 | 92 | |||
AK212606.1 | - | 1 | 92 | |||
AK214260.1 | - | 1 | 92 | |||
AK214872.1 | - | 1 | 92 | |||
AK215152.1 | - | 1 | 92 | |||
BC011058.1 | - | 3 | 780 | |||
BC052378.1 | - | 7 | 2476 | 5696275 | ||
BC054447.1 | - | 7 | 2468 | 6477698 |
