siRNA Details
Gene Symbol: Eno1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023119.2 | NP_075608.2 | 7 | 722 | ||
GenBank mRNA | AK002322.1 | - | 7 | 681 | ||
AK002336.1 | - | 7 | 680 | |||
AK088788.1 | - | 7 | 666 | |||
AK147987.1 | - | 7 | 680 | |||
AK150161.1 | - | 7 | 680 | |||
AK150192.1 | - | 7 | 681 | |||
AK153321.1 | - | 7 | 679 | |||
AK202509.1 | - | 4 | 382 | |||
BC003891.1 | - | 7 | 672 | 3592683 | ||
BC003934.1 | - | 7 | 694 | |||
BC010685.1 | - | 7 | 660 | 3599051 | ||
BC024644.1 | - | 7 | 652 | 3583123 | ||
BC039179.1 | - | 7 | 642 | 4501041 | ||
BC056611.1 | - | 3 | 848 | 4948983 | ||
BC083334.1 | - | 7 | 654 | 6414729 | ||
BC085098.1 | - | 6 | 583 | 5376359 | ||
X52379.1 | - | 7 | 664 |
Gene Symbol: Eno1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025388.1 | NP_001020559.1 | 2 | 704 | ||
GenBank mRNA | BC004017.1 | - | 1 | 326 | ||
BC089539.1 | - | 2 | 704 | 6707317 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
