siRNA Details
Gene Symbol: Pla2g12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286948.1 | NP_001273877.1 | 4 | 1150 | ||
NM_023196.4 | NP_075685.2 | 4 | 1081 | |||
NM_183423.3 | NP_904359.1 | 4 | 864 | |||
GenBank mRNA | AK009133.1 | - | 4 | 1083 | ||
AK163693.1 | - | 4 | 995 | |||
AY007381.1 | - | 4 | 1057 | |||
BC051117.1 | - | 4 | 1081 | 6306803 | ||
BC055051.1 | - | 1 | 2516 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
