siRNA Details
Gene Symbol: Ndnl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF319979.1 | - | 1 | 227 | ||
AK010294.1 | - | 1 | 310 | |||
AK018250.1 | - | 1 | 310 | |||
AK049759.1 | - | 1 | 329 | |||
AK076471.1 | - | 1 | 335 | |||
AK131157.1 | - | 306 | ||||
BC034892.1 | - | 1 | 274 | 1494952 | ||
BC092289.1 | - | 1 | 311 | 4990134 |

More Information
Other Species Targeted

