siRNA Details
Gene Symbol: Krt42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212483.2 | NP_997648.2 | 6 | 1123 | ||
GenBank mRNA | AK019115.1 | - | 3 | 640 | ||
AK019133.1 | - | 3 | 585 | |||
AY459292.1 | - | 6 | 1123 | |||
BC112371.1 | - | 6 | 1115 | |||
BC148204.1 | - | 6 | 1123 | 40056537 |

More Information
Other Species Targeted

Gene Ontology Categories:
