siRNA Details
Gene Symbol: 2010204K13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF038507.1 | - | 2 | 471 | ||
AK002502.1 | - | 856 | ||||
AK008437.1 | - | 2 | 508 | |||
AK077356.1 | - | 2 | 551 | |||
BC042200.1 | - | 2 | 470 | |||
BC061056.1 | - | 2 | 518 | |||
BC072610.1 | - | 2 | 431 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
